missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Hydroxyphenyl)glycine 97.0+%, TCI America™

Catalog No. H029225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H029225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H029225G Supplier TCI America Supplier No. H029225G
Only null left

Chemical Identifiers

CAS 122-87-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00002689
InChI Key WRUZLCLJULHLEY-UHFFFAOYSA-N
Synonym n-4-hydroxyphenyl glycine, photoglycine, p-hydroxyphenylglycine, hydroxyphenyl glycine, p-hydroxyanilinoacetic acid, iconyl, p-hydroxyphenyl glycine, glycine, n-4-hydroxyphenyl, 4-carboxymethylamino phenol, glitsin
PubChem CID 67149
ChEBI CHEBI:55443
IUPAC Name 2-(4-hydroxyanilino)acetic acid
SMILES C1=CC(=CC=C1NCC(=O)O)O

Specifications

Quantity 25 g
Formula Weight 167.16
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Hydroxyphenyl)glycine
EINECSNumber (3)-0681
TSCA Yes