missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorophenyl)glycine 98.0+%, TCI America™

Catalog No. F09601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09601G 1 g
F09605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09601G Supplier TCI America Supplier No. F09601G
Only null left

Chemical Identifiers

CAS 351-95-1
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD02255629
InChI Key ZFIMCCCQJDZLCT-UHFFFAOYSA-N
PubChem CID 1133296
IUPAC Name 2-[(4-fluorophenyl)amino]acetic acid
SMILES OC(=O)CNC1=CC=C(F)C=C1

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 1 g
Formula Weight 169.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Fluorophenyl)glycine