missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog No. aah56745md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH56745MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH56745MD Supplier Thermo Scientific Chemicals Supplier No. H56745MD
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1041583-33-8
Molecular Formula C14H11F4N
Molecular Weight (g/mol) 269.24
MDL Number MFCD12618204
InChI Key JCWUQELJSDRKEP-UHFFFAOYSA-N
Synonym n-4-fluorobenzyl-2-trifluoromethyl aniline, n-4-fluorophenyl methyl-2-trifluoromethyl aniline
PubChem CID 29292080
IUPAC Name N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline
SMILES FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1

Specifications

Quantity 250 mg
Formula Weight 269.24
Percent Purity 97%
Chemical Name or Material N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures