missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Chlorophenyl)phthalimide 98.0+%, TCI America™
$161.72 - $758.52
Chemical Identifiers
| CAS | 7386-21-2 |
|---|---|
| Molecular Formula | C14H8ClNO2 |
| Molecular Weight (g/mol) | 257.67 |
| MDL Number | MFCD00023048 |
| InChI Key | QKHKQJWODBAIMN-UHFFFAOYSA-N |
| Synonym | n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide |
| PubChem CID | 81867 |
| IUPAC Name | 2-(4-chlorophenyl)-2,3-dihydro-1H-isoindole-1,3-dione |
| SMILES | ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O |
Chemical Identifiers
| 7386-21-2 | |
| 257.67 | |
| QKHKQJWODBAIMN-UHFFFAOYSA-N | |
| 81867 | |
| ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O |
| C14H8ClNO2 | |
| MFCD00023048 | |
| n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide | |
| 2-(4-chlorophenyl)-2,3-dihydro-1H-isoindole-1,3-dione |
Specifications
| 7386-21-2 | |
| White | |
| MFCD00023048 | |
| n-4-chlorophenyl phthalimide, n-p-chlorophenyl phthalimide, phthalimide, n-p-chlorophenyl, 1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl, 2-4-chlorophenyl isoindoline-1,3-dione, 2-4-chlorophenyl-1h-isoindole-1,3 2h-dione, 2-4-chlorophenyl isoindole-1,3-dione, acmc-209osn, 5-21-10-00293 beilstein handbook reference, n-4-chlorophenyl-phthalimide | |
| ClC1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O | |
| 257.67 | |
| 257.67 | |
| Crystalline Powder |
| 198°C | |
| C14H8ClNO2 | |
| 5 g | |
| QKHKQJWODBAIMN-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)-2,3-dihydro-1H-isoindole-1,3-dione | |
| 81867 | |
| ≥98.0% (GC) | |
| N-(4-Chlorophenyl)phthalimide |
Safety and Handling
RTECSNumber : TI4267000
TSCA : No