missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Chloro-2-butynyl)phthalimide, 97%

CAS: 4819-69-6 | C12H8ClNO2 | 233.651 g/mol

$158.71 - $570.19

Chemical Identifiers

CAS 4819-69-6
Molecular Formula C12H8ClNO2
Molecular Weight (g/mol) 233.651
MDL Number MFCD01318120
InChI Key TXNDRPKNOXQAAO-UHFFFAOYSA-N
Synonym n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
PubChem CID 237537
IUPAC Name 2-(4-chlorobut-2-ynyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1631303
View Documents
Thermo Scientific Chemicals
L1631303
1 g
Each for $158.71
Only null left
 
AAL1631306
View Documents
Thermo Scientific Chemicals
L1631306
5 g
Each for $570.19
Only null left
 
Description

Description

N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

4819-69-6
233.651
TXNDRPKNOXQAAO-UHFFFAOYSA-N
237537
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl
C12H8ClNO2
MFCD01318120
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione

Specifications

4819-69-6
Nearly Odorless
MFCD01318120
UN3261
n-4-chloro-2-butynyl phthalimide, 2-4-chlorobut-2-yn-1-yl isoindoline-1,3-dione, 2-4-chlorobut-2-yn-1-yl-1h-isoindole-1,3 2h-dione, 2-4-chlorobut-2-yn-1-yl isoindole-1,3-dione, acmc-20akbi, n-4-chlorobut-2-ynyl phthalimide, 2-4-chlorobut-2-ynyl isoindole-1,3-dione, 2-4-chloro-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-4-chloro-2-butyn-1-yl, 2-4-chlorobut-2-yn-1-yl-2,3-dihydro-1h-isoindole-1,3-dione
TXNDRPKNOXQAAO-UHFFFAOYSA-N
2-(4-chlorobut-2-ynyl)isoindole-1,3-dione
237537
97%
115°C to 120°C
C12H8ClNO2
1 g
179626
Insoluble in water.
C1=CC=C2C(=C1)C(=O)N(C2=O)CC#CCCl
233.651
233.66
N-(4-Chloro-2-butynyl)phthalimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only