missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromophenyl)maleimide, 98%

Catalog No. aaa1311914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1311914 25 g
AAA1311903 1 g
AAA1311906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1311914 Supplier Thermo Scientific Chemicals Supplier No. A1311914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13380-67-1 | C10H6BrNO2 | 252.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13380-67-1
Molecular Formula C10H6BrNO2
Molecular Weight (g/mol) 252.067
MDL Number MFCD00030664
InChI Key FECSFBYOMHWJQG-UHFFFAOYSA-N
Synonym n-4-bromophenyl maleimide, 1-4-bromophenyl-1h-pyrrole-2,5-dione, 1-4-bromophenyl pyrrole-2,5-dione, 1-4-bromophenyl-2,5-dioxo-pyrrole, 1-4-bromophenyl maleimide, 1-4-bromophenyl-2,5-dihydro-1h-pyrrole-2,5-dione, 1-4-bromphenyl-1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione,1-4-bromophenyl, 1h-pyrrole-2,5-dione, 1-4-bromophenyl, 1-4-bromophenyl azoline-2,5-dione
PubChem CID 123301
IUPAC Name 1-(4-bromophenyl)pyrrole-2,5-dione
SMILES C1=CC(=CC=C1N2C(=O)C=CC2=O)Br

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Beilstein 513122
Formula Weight 252.07
Percent Purity 98%
Chemical Name or Material N-(4-Bromophenyl)maleimide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only