missing translation for 'onlineSavingsMsg'
Learn More

N-(4'-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine 98.0+%, TCI America™

Catalog No. p-7123918
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5049200MG 200 mg
B50491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5049200MG Supplier TCI America Supplier No. B5049200MG
Only null left

Chemical Identifiers

CAS 352359-42-3
Molecular Formula C28H20BrN
Molecular Weight (g/mol) 450.379
MDL Number MFCD24387457
InChI Key JYAVHXABZQYLTH-UHFFFAOYSA-N
Synonym N-(4′C-Bromo-[1,1′C-biphenyl]-4-yl)-N-phenylnaphthalen-1-amine
PubChem CID 16755312
IUPAC Name N-[4-(4-bromophenyl)phenyl]-N-phenylnaphthalen-1-amine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)C3=CC=C(C=C3)Br)C4=CC=CC5=CC=CC=C54

Specifications

Melting Point 169°C
Color White-Yellow
Quantity 200 mg
Formula Weight 450.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4′-Bromo-4-biphenylyl)-N-phenyl-1-naphthylamine