missing translation for 'onlineSavingsMsg'
Learn More

N'-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine 98.0+%, TCI America™

Catalog No. p-7123492
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43621G 1 g
B43625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43621G Supplier TCI America Supplier No. B43621G
Only null left

Chemical Identifiers

CAS 39255-60-2
Molecular Formula C10H10BrN3
Molecular Weight (g/mol) 252.12
MDL Number MFCD27950293
InChI Key ZGEYANFICVYJSY-UHFFFAOYSA-N
PubChem CID 57857968
IUPAC Name N'-(4-bromo-2-cyanophenyl)-N,N-dimethylmethanimidamide
SMILES CN(C)C=NC1=C(C=C(Br)C=C1)C#N

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 1 g
Synonym N′C-(4-Bromo-2-cyanophenyl)-N,N-dimethylformimidamide
Formula Weight 252.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N′-(4-Bromo-2-cyanophenyl)-N,N-dimethylformamidine