missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Biphenylyl)-2-biphenylamine 98.0+%, TCI America™
$130.48 - $460.43
Chemical Identifiers
| CAS | 1372775-52-4 |
|---|---|
| Molecular Formula | C24H19N |
| Molecular Weight (g/mol) | 321.423 |
| MDL Number | MFCD22988894 |
| InChI Key | LIBHEMBTFRBMOV-UHFFFAOYSA-N |
| PubChem CID | 86762387 |
| IUPAC Name | 2-phenyl-N-(4-phenylphenyl)aniline |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3C4=CC=CC=C4 |
Chemical Identifiers
| 1372775-52-4 | |
| 321.423 | |
| LIBHEMBTFRBMOV-UHFFFAOYSA-N | |
| 2-phenyl-N-(4-phenylphenyl)aniline |
| C24H19N | |
| MFCD22988894 | |
| 86762387 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3C4=CC=CC=C4 |
Specifications
| 1372775-52-4 | |
| White | |
| MFCD22988894 | |
| LIBHEMBTFRBMOV-UHFFFAOYSA-N | |
| 2-phenyl-N-(4-phenylphenyl)aniline | |
| 86762387 | |
| ≥98.0% (GC) | |
| N-(4-Biphenylyl)-2-biphenylamine |
| 111°C | |
| C24H19N | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3C4=CC=CC=C4 | |
| 321.423 | |
| 321.42 | |
| Crystalline Powder |
Safety and Handling
TSCA : No