missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Biphenylyl)-2-biphenylamine 98.0+%, TCI America™

Catalog No. B48665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48665G 5 g
B48661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48665G Supplier TCI America Supplier No. B48665G
Only null left

Chemical Identifiers

CAS 1372775-52-4
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.423
MDL Number MFCD22988894
InChI Key LIBHEMBTFRBMOV-UHFFFAOYSA-N
PubChem CID 86762387
IUPAC Name 2-phenyl-N-(4-phenylphenyl)aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=CC=C3C4=CC=CC=C4

Specifications

Melting Point 111°C
Color White
Quantity 5 g
Formula Weight 321.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Biphenylyl)-2-biphenylamine