missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminophenyl)maleimide 93.0+%, TCI America™

Catalog No. A0825100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0825100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0825100MG Supplier TCI America Supplier No. A0825100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29753-26-2
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00051191
InChI Key XOPCHXSYQHXLHJ-UHFFFAOYSA-N
PubChem CID 122469
IUPAC Name 1-(4-aminophenyl)pyrrole-2,5-dione
SMILES C1=CC(=CC=C1N)N2C(=O)C=CC2=O

Specifications

Melting Point 173°C
Quantity 100 mg
Formula Weight 188.19
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminophenyl)maleimide