missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzoyl)-L-glutamic Acid 97.0+%, TCI America™

Catalog No. A04425G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04425G 5 g
A04421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04425G Supplier TCI America Supplier No. A04425G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4271-30-1
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.253
MDL Number MFCD00042821
InChI Key GADGMZDHLQLZRI-VIFPVBQESA-N
PubChem CID 196473
ChEBI CHEBI:61114
IUPAC Name (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)N

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 5 g
Formula Weight 266.25
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic Acid
EINECSNumber (9)-1580
TSCA No