missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzoyl)-beta-alanine 98.0+%, TCI America™

Catalog No. A23475G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23475G 5 g
A234725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23475G Supplier TCI America Supplier No. A23475G

Chemical Identifiers

CAS 7377-08-4
Molecular Formula C10H11N2O3
Molecular Weight (g/mol) 207.21
MDL Number MFCD00009805
InChI Key VHAXWROFYVPXMZ-UHFFFAOYSA-M
Synonym n-4-aminobenzoyl-beta-alanine, 3-4-amino-benzoylamino-propionic acid, 3-4-aminobenzamido propanoic acid, 4-aminobenzoyl-beta-alanine, unii-ab85ljq945, 3-4-aminobenzoyl amino propanoic acid, n-4-aminophenyl carbonyl-beta-alanine, 3-4-aminophenyl formamido propanoic acid, 3-4-aminophenyl carbonylamino propanoic acid
PubChem CID 719629
IUPAC Name 3-[(4-aminophenyl)formamido]propanoate
SMILES NC1=CC=C(C=C1)C(=O)NCCC([O-])=O

Specifications

Melting Point 155°C
Color Yellow
Quantity 5 g
Formula Weight 208.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Aminobenzoyl)-beta-alanine