missing translation for 'onlineSavingsMsg'
Learn More

N(4)-Acetylcytosine, 98%

CAS: 14631-20-0 | C6H7N3O2 | 153.14 g/mol

Supplier:  Thermo Scientific Chemicals B2085114

 View more versions of this product

Catalog No. AAB2085114


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N(4)-Acetylcytosine
>300°C
MFCD00134466
n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone
CC(=O)NC1=CC=NC(=O)N1
153.14
153.14
14631-20-0
C6H7N3O2
25 g
IJCKBIINTQEGLY-UHFFFAOYSA-N
N-(2-oxo-1H-pyrimidin-6-yl)acetamide
99309
98%

Chemical Identifiers

14631-20-0
153.14
IJCKBIINTQEGLY-UHFFFAOYSA-N
N-(2-oxo-1H-pyrimidin-6-yl)acetamide
C6H7N3O2
MFCD00134466
99309
CC(=O)NC1=CC=NC(=O)N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only