missing translation for 'onlineSavingsMsg'
Learn More

N(4)-Acetylcytosine, 98%

Catalog No. AAB2085106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2085106 5 g
AAB2085114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2085106 Supplier Thermo Scientific Chemicals Supplier No. B2085106

CAS: 14631-20-0 | C6H7N3O2 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14631-20-0
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00134466
InChI Key IJCKBIINTQEGLY-UHFFFAOYSA-N
PubChem CID 99309
IUPAC Name N-(2-oxo-1H-pyrimidin-6-yl)acetamide
SMILES CC(=O)NC1=CC=NC(=O)N1

Specifications

Melting Point >300°C
Quantity 5 g
Synonym n4-acetylcytosine, n-acetylcytosine, n-2-oxo-1,2-dihydropyrimidin-4-yl acetamide, acetamide, n-1,2-dihydro-2-oxo-4-pyrimidinyl, n 4-acetylcytosine, n-2-oxo-1h-pyrimidin-6-yl acetamide, n-2-hydroxypyrimidin-4-yl acetamide, n-2-oxo-3h-pyrimidin-4-yl acetamide, 4-acetylamino-1,2-dihydro-2-pyrimidone
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material N(4)-Acetylcytosine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only