missing translation for 'onlineSavingsMsg'
Learn More

N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide, 98%

CAS: 147118-37-4 | C16H18FN3O3S | 351.396 g/mol

$188.89 - $554.30

Chemical Identifiers

CAS 147118-37-4
Molecular Formula C16H18FN3O3S
Molecular Weight (g/mol) 351.396
MDL Number MFCD08458342
InChI Key WOCOTUDOVSLFOB-UHFFFAOYSA-N
Synonym n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
PubChem CID 10473133
IUPAC Name N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
SMILES CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6112914
View Documents
Thermo Scientific Chemicals
H6112914
25 g
Each for $188.89
Only null left
 
AAH6112922
View Documents
Thermo Scientific Chemicals
H6112922
100 g
Each for $554.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

147118-37-4
351.396
WOCOTUDOVSLFOB-UHFFFAOYSA-N
10473133
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
C16H18FN3O3S
MFCD08458342
n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide

Specifications

147118-37-4
C16H18FN3O3S
25 g
WOCOTUDOVSLFOB-UHFFFAOYSA-N
N-[4-(4-fluorophenyl)-5-formyl-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
10473133
99%
N-[4-(4-Fluorophenyl)-5-formyl-6-isopropyl-2-pyrimidinyl]-N-methylmethanesulfonamide
White
MFCD08458342
n-4-4-fluorophenyl-5-formyl-6-isopropylpyrimidin-2-yl-n-methylmethanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonyl amino pyrimidinyl-5-yl-formyl, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino-5-pyrimidinecarboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carboxaldehyde, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methanesulfonylamino pyrimidin-5-carboxaldehyde, n-4-4-fluorophenyl-5-formyl-6-isopropyl-pyrimidin-2-yl-n-methyl-methanesulfonamide, n-4-4-fluorophenyl-5-formyl-6-1-methylethyl-2-pyrimidinyl-n-methyl-methanesulfonamide, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-met, 4-4-fluorophenyl-6-isopropyl-2-n-methyl-n-methylsulfonylamino pyrimidine-5-carbaldehyde
CC(C)C1=NC(=NC(=C1C=O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C
351.396
351.4
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only