Learn More
N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, 50% in methanol
CAS: 35141-36-7 | C9H24ClNO3Si | 257.83 g/mol
$60.01 - $637.27
Chemical Identifiers
| CAS | 35141-36-7 |
|---|---|
| Molecular Formula | C9H24ClNO3Si |
| Molecular Weight (g/mol) | 257.83 |
| MDL Number | MFCD00054225 |
| InChI Key | FYZFRYWTMMVDLR-UHFFFAOYSA-M |
| Synonym | n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium |
| PubChem CID | 94462 |
| IUPAC Name | trimethyl(3-trimethoxysilylpropyl)azanium;chloride |
| SMILES | C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6641414
|
Thermo Scientific Chemicals
H6641414 |
25 g |
Each for $60.01
|
|
|||||
|
AAH6641422
|
Thermo Scientific Chemicals
H6641422 |
100 g |
Each for $149.70
|
|
|||||
|
AAH6641436
|
Thermo Scientific Chemicals
H6641436 |
500 g |
Each for $637.27
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 35141-36-7 | |
| 257.83 | |
| FYZFRYWTMMVDLR-UHFFFAOYSA-M | |
| 94462 | |
| C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] |
| C9H24ClNO3Si | |
| MFCD00054225 | |
| n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium | |
| trimethyl(3-trimethoxysilylpropyl)azanium;chloride |
Specifications
| 35141-36-7 | |
| 0.930 g/mL | |
| C9H24ClNO3Si | |
| MFCD00054225 | |
| UN1230 | |
| Hygroscopic | |
| FYZFRYWTMMVDLR-UHFFFAOYSA-M | |
| trimethyl(3-trimethoxysilylpropyl)azanium;chloride | |
| 94462 | |
| Liquid |
| Colorless to Yellow | |
| 11°C (52°F) | |
| 1.397 | |
| 25 g | |
| 11593127 | |
| n-trimethoxysilylpropyl-n,n,n-trimethylammonium chloride, n,n,n-trimethyl-3-trimethoxysilyl propan-1-aminium chloride, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride, trimethyl-3-trimethoxysilyl propylammonium chloride, trimethyl 3-trimethoxysilyl propyl azanium chloride, 1-propanaminium,n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 1-propanaminium, n,n,n-trimethyl-3-trimethoxysilyl-, chloride 1:1, 3-trimethoxysilylpropyl trimethylammonium chloride, acmc-20ak0r, trimethyl-3-trimethoxysilyl propyl ammonium | |
| C[N+](C)(C)CCC[Si](OC)(OC)OC.[Cl-] | |
| 257.83 | |
| 257.83 | |
| N-[3-(Trimethoxysilyl)propyl]-N,N,N-trimethylammonium chloride, in 50% methanol |
Safety and Handling
GHS H Statement
H225-H311-H331-H370-H335-H302-H315-H319
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
H225-H301+H311+H331-H370
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: METHANOL
EINECSNumber : 252-393-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only