missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxypropyl)ethylenediamine 97.0+%, TCI America™

$348.46 - $1386.13

Chemical Identifiers

CAS 56344-32-2
Molecular Formula C5H14N2O
Molecular Weight (g/mol) 118.18
MDL Number MFCD00059848
InChI Key KTLIZDDPOZZHCD-UHFFFAOYSA-N
Synonym N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine
PubChem CID 10973740
IUPAC Name 3-(2-aminoethylamino)propan-1-ol
SMILES C(CNCCN)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H03915G
View Documents
TCI America
H03915G
5 g
Each for $348.46
Only null left
 
H039125G
View Documents
TCI America
H039125G
25 g
Each for $1,386.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56344-32-2
118.18
KTLIZDDPOZZHCD-UHFFFAOYSA-N
10973740
C(CNCCN)CO
C5H14N2O
MFCD00059848
N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine
3-(2-aminoethylamino)propan-1-ol

Specifications

56344-32-2
White-Yellow
C5H14N2O
5 g
N-(2-Aminoethyl)propanolamine, 2-(3-Hydroxypropylamino)ethylamine
C(CNCCN)CO
118.18
118.18
Crystalline Lumps
49°C
149°C
MFCD00059848
3259
KTLIZDDPOZZHCD-UHFFFAOYSA-N
3-(2-aminoethylamino)propan-1-ol
10973740
≥97.0% (GC,T)
N-(3-Hydroxypropyl)ethylenediamine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents