missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Hydroxypropyl)ethylenediamine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 49°C |
| Color | White-Yellow |
| Boiling Point | 149°C |
| Quantity | 5 g |
| UN Number | 3259 |
| Formula Weight | 118.18 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | N-(3-Hydroxypropyl)ethylenediamine |
Safety and Handling
| TSCA | No |