missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Dimethylaminopropyl)methacrylamide (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. D191925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191925G 25 g
D1919100G 100 g
D1919500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D191925G Supplier TCI America Supplier No. D191925G

Chemical Identifiers

CAS 5205-93-6
Molecular Formula C9H18N2O
Molecular Weight (g/mol) 170.256
MDL Number MFCD00038359
InChI Key GDFCSMCGLZFNFY-UHFFFAOYSA-N
Synonym n-3-dimethylamino propyl methacrylamide, dmapma, unii-63obq7bzds, 63obq7bzds, n-dimethylamino propyl methacrylamide, n-3-dimethylaminopropyl methacrylamide, n-3-dimethylamino propyl-2-methylprop-2-enamide, 2-propenamide, n-3-dimethylamino propyl-2-methyl
PubChem CID 78882
IUPAC Name N-[3-(dimethylamino)propyl]-2-methylprop-2-enamide
SMILES CC(=C)C(=O)NCCCN(C)C

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 170.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(3-Dimethylaminopropyl)methacrylamide (stabilized with MEHQ)
EINECSNumber (2)-3195
TSCA Yes