missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ) 98.0+%, TCI America™
Supplier: TCI America D1785100G
Specifications
| N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ) | |
| Yellow | |
| C8H16N2O | |
| 100 g | |
| CN(C)CCCNC(=O)C=C | |
| 156.23 | |
| 156.23 | |
| Liquid |
| 3845-76-9 | |
| 142°C | |
| MFCD00059902 | |
| ADTJPOBHAXXXFS-UHFFFAOYSA-N | |
| N-[3-(dimethylamino)propyl]prop-2-enamide | |
| 77452 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 3845-76-9 | |
| 156.23 | |
| ADTJPOBHAXXXFS-UHFFFAOYSA-N | |
| N-[3-(dimethylamino)propyl]prop-2-enamide |
| C8H16N2O | |
| MFCD00059902 | |
| 77452 | |
| CN(C)CCCNC(=O)C=C |
Safety and Handling
EINECSNumber : (2)-1013
TSCA : No
Recommended Storage : Refrigerator