missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. D178525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D178525G 25 g
D1785100G 100 g
D1785500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D178525G Supplier TCI America Supplier No. D178525G
Only null left

Chemical Identifiers

CAS 3845-76-9
Molecular Formula C8H16N2O
Molecular Weight (g/mol) 156.23
MDL Number MFCD00059902
InChI Key ADTJPOBHAXXXFS-UHFFFAOYSA-N
PubChem CID 77452
IUPAC Name N-[3-(dimethylamino)propyl]prop-2-enamide
SMILES CN(C)CCCNC(=O)C=C

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 g
Formula Weight 156.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[3-(Dimethylamino)propyl]acrylamide (stabilized with MEHQ)
EINECSNumber (2)-1013
TSCA No
Recommended Storage Refrigerator