missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Azidopropyl)biotinamide 95.0+%, TCI America™

Catalog No. a2524100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2524100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2524100MG Supplier TCI America Supplier No. A2524100MG

Chemical Identifiers

CAS 908007-17-0
Molecular Formula C13H22N6O2S
Molecular Weight (g/mol) 326.419
MDL Number MFCD20134144
InChI Key SWODDJWJUGOAQB-NHCYSSNCSA-N
PubChem CID 59828156
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-(3-azidopropyl)pentanamide
SMILES C1C2C(C(S1)CCCCC(=O)NCCCN=[N+]=[N-])NC(=O)N2

Specifications

Melting Point 168°C
Color White
Quantity 100 mg
Formula Weight 326.42
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Azidopropyl)biotinamide