missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)-N-methylaniline 96.0+%, TCI America™

Catalog No. p-7117354
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A129525ML 25 mL
A1295500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A129525ML Supplier TCI America Supplier No. A129525ML

Chemical Identifiers

CAS 53485-07-7
Molecular Formula C10H16N2
Molecular Weight (g/mol) 164.25
MDL Number MFCD00191602
InChI Key WGHBKYWUOOANEX-UHFFFAOYSA-N
PubChem CID 577774
IUPAC Name N-(3-aminopropyl)-N-methylaniline
SMILES CN(CCCN)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 mL
Formula Weight 164.25
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(3-Aminopropyl)-N-methylaniline