missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Aminopropyl)morpholine 99.0+%, TCI America™

Catalog No. A040925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A040925ML 25 mL
A0409500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A040925ML Supplier TCI America Supplier No. A040925ML

Chemical Identifiers

CAS 123-00-2
Molecular Formula C7H16N2O
Molecular Weight (g/mol) 144.22
MDL Number MFCD00006184
InChI Key UIKUBYKUYUSRSM-UHFFFAOYSA-N
Synonym n-3-aminopropyl morpholine, 4-morpholinepropanamine, 3-morpholinopropylamine, 3-morpholinopropan-1-amine, n-aminopropylmorpholine, 4-3-aminopropyl morpholine, 4-morpholinepropylamine, 4-aminopropylmorpholine, 1-amino-3-morpholinopropane, 3-4-morpholinyl-1-propanamine
PubChem CID 61055
ChEBI CHEBI:86554
IUPAC Name 3-(morpholin-4-yl)propan-1-amine
SMILES NCCCN1CCOCC1

Specifications

Color Yellow
Boiling Point 225°C
Quantity 25 mL
UN Number 2735
Formula Weight 144.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(3-Aminopropyl)morpholine
EINECSNumber (5)-0862
RTECSNumber QD7700000
TSCA Yes