missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Aminopropyl)diethanolamine 90.0+%, TCI America™
$205.05 - $1672.06
Chemical Identifiers
| CAS | 4985-85-7 |
|---|---|
| Molecular Formula | C7H18N2O2 |
| Molecular Weight (g/mol) | 162.23 |
| MDL Number | MFCD00047971 |
| InChI Key | FKJVYOFPTRGCSP-UHFFFAOYSA-N |
| Synonym | N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane |
| PubChem CID | 21088 |
| IUPAC Name | 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethan-1-ol |
| SMILES | NCCCN(CCO)CCO |
Chemical Identifiers
| 4985-85-7 | |
| 162.23 | |
| FKJVYOFPTRGCSP-UHFFFAOYSA-N | |
| 21088 | |
| NCCCN(CCO)CCO |
| C7H18N2O2 | |
| MFCD00047971 | |
| N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane | |
| 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethan-1-ol |
Specifications
| 4985-85-7 | |
| 12 | |
| C7H18N2O2 | |
| 25 g | |
| N,N-Bis(2-hydroxyethyl)-1,3-diaminopropane | |
| NCCCN(CCO)CCO | |
| 162.23 | |
| 162.23 | |
| Liquid |
| Yellow | |
| 207°C | |
| MFCD00047971 | |
| 2735 | |
| FKJVYOFPTRGCSP-UHFFFAOYSA-N | |
| 2-[(3-aminopropyl)(2-hydroxyethyl)amino]ethan-1-ol | |
| 21088 | |
| ≥90.0% (GC) | |
| N-(3-Aminopropyl)diethanolamine |
Safety and Handling
TSCA : Yes