missing translation for 'onlineSavingsMsg'
Learn More

N-(3'-Aminopropyl)-2-pyrrolidinone, 95%

Catalog No. AC159600050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159600050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159600050 Supplier Thermo Scientific Chemicals Supplier No. 159600050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7663-77-6 | C7H14N2O | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7663-77-6
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.20
MDL Number MFCD00003201
InChI Key HJORCZCMNWLHMB-UHFFFAOYSA-N
Synonym 1-3-aminopropyl pyrrolidin-2-one, n-3'-aminopropyl-2-pyrrolidinone, 2-pyrrolidinone, 1-3-aminopropyl, 1-3-aminopropyl-2-pyrrolidone, n-3-aminopropyl-2-pyrrolidinone, 1-3-aminopropyl-2-pyrrolidinone, n-3-aminopropyl pyrrolidin-2-one, 1-3-amino-propyl-pyrrolidin-2-one, 1-3-aminoprop-1-yl pyrrolidin-2-one, zlchem 943
PubChem CID 82111
IUPAC Name 1-(3-aminopropyl)pyrrolidin-2-one
SMILES NCCCN1CCCC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.0100g/mL
Boiling Point 120.0°C to 123.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 1,5-Diazabicyclo(4.3.0.)non-5-ene: Balance (GC)
Packaging Glass bottle
Refractive Index 1.4970 to 1.4990
Quantity 5 g
Specific Gravity 1.01
Formula Weight 142.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-(3'-Aminopropyl)-2-pyrrolidinone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
EINECSNumber 231-632-7

RUO – Research Use Only