missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(2-Furyl)benzyl]-N-methylamine, ≥97%, Thermo Scientific™

Catalog No. CC46146DA
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC46146DA 1g
CC46146CB 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC46146DA Supplier Thermo Scientific Chemicals Supplier No. CC46146DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 857284-27-6
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.242
MDL Number MFCD08435877
InChI Key WMCXJYGZLCREMM-UHFFFAOYSA-N
Synonym n-3-2-furyl benzyl-n-methylamine, 3-furan-2-yl phenyl methyl methyl amine, 1-3-furan-2-yl phenyl-n-methylmethanamine, 3-2-furyl phenyl methyl methylamine, benzenemethanamine,3-2-furanyl-n-methyl
PubChem CID 18525790
IUPAC Name 1-[3-(furan-2-yl)phenyl]-N-methylmethanamine
SMILES CNCC1=CC=CC(=C1)C2=CC=CO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1g
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material N-[3-(2-furyl)benzyl]-N-methylamine
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.