missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Propynyl)succinimide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 53°C |
| Color | White-Yellow |
| Boiling Point | 237°C |
| Quantity | 1 g |
| Formula Weight | 137.14 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(2-Propynyl)succinimide |
Safety and Handling
| TSCA | No |