missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Nitrophenylsulfenyl)-S-benzyl-L-cysteine Dicyclohexylammonium Salt 99.0+%, TCI America™
$92.27 - $236.56
Chemical Identifiers
| CAS | 7675-65-2 |
|---|---|
| Molecular Formula | C28H39N3O4S2 |
| Molecular Weight (g/mol) | 545.76 |
| MDL Number | MFCD00038490 |
| InChI Key | MAUIFYQGIWFDHJ-UHFFFAOYNA-N |
| Synonym | N-(2-Nitrophenylthio)-S-benzyl-L-cysteine Dicyclohexylammonium Salt, N-Nps-S-benzyl-L-cysteine Dicyclohexylammonium Salt, Nps-Cys(Bzl)-OH.DCHA |
Chemical Identifiers
| 7675-65-2 | |
| 545.76 | |
| MAUIFYQGIWFDHJ-UHFFFAOYNA-N |
| C28H39N3O4S2 | |
| MFCD00038490 | |
| N-(2-Nitrophenylthio)-S-benzyl-L-cysteine Dicyclohexylammonium Salt, N-Nps-S-benzyl-L-cysteine Dicyclohexylammonium Salt, Nps-Cys(Bzl)-OH.DCHA |
Specifications
| 7675-65-2 | |
| C28H39N3O4S2 | |
| 1 g | |
| MAUIFYQGIWFDHJ-UHFFFAOYNA-N | |
| 545.76 | |
| Crystalline Powder |
| Yellow | |
| MFCD00038490 | |
| N-(2-Nitrophenylthio)-S-benzyl-L-cysteine Dicyclohexylammonium Salt, N-Nps-S-benzyl-L-cysteine Dicyclohexylammonium Salt, Nps-Cys(Bzl)-OH.DCHA | |
| 545.76 | |
| ≥99.0% (T) | |
| N-(2-Nitrophenylsulfenyl)-S-benzyl-L-cysteine Dicyclohexylammonium Salt |
Safety and Handling
TSCA : No