missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Nitrophenylsulfenyl)-S-benzyl-L-cysteine Dicyclohexylammonium Salt 99.0+%, TCI America™

Catalog No. N03615G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03615G 5 g
N03611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03615G Supplier TCI America Supplier No. N03615G

Chemical Identifiers

CAS 7675-65-2
Molecular Formula C28H39N3O4S2
Molecular Weight (g/mol) 545.76
MDL Number MFCD00038490
InChI Key MAUIFYQGIWFDHJ-UHFFFAOYNA-N
Synonym N-(2-Nitrophenylthio)-S-benzyl-L-cysteine Dicyclohexylammonium Salt, N-Nps-S-benzyl-L-cysteine Dicyclohexylammonium Salt, Nps-Cys(Bzl)-OH.DCHA

Specifications

Color Yellow
Quantity 5 g
Formula Weight 545.76
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Nitrophenylsulfenyl)-S-benzyl-L-cysteine Dicyclohexylammonium Salt