missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-2-Nitrophenylsulfenyl-L-alanine Dicyclohexylammonium Salt 98.0+%, TCI America™
Supplier: TCI America N04141G
Specifications
| N-2-Nitrophenylsulfenyl-L-alanine Dicyclohexylammonium Salt | |
| 176°C | |
| C21H33N3O4S | |
| 1 g | |
| LGTGUPPUIQQLBP-ZCMDIHMWSA-N | |
| N-cyclohexylcyclohexanamine;(2S)-2-[(2-nitrophenyl)sulfanylamino]propanoic acid | |
| 56842015 | |
| ≥98.0% (HPLC) |
| 7675-46-9 | |
| Yellow | |
| MFCD00066085 | |
| N-(2-Nitrophenylthio)-L-alanine Dicyclohexylammonium Salt, N-Nps-L-alanine Dicyclohexylammonium Salt, Nps-Ala-OH.DCHA | |
| CC(C(=O)O)NSC1=CC=CC=C1[N+](=O)[O-].C1CCC(CC1)NC2CCCCC2 | |
| 423.572 | |
| 423.57 | |
| Crystalline Powder |
Chemical Identifiers
| 7675-46-9 | |
| 423.572 | |
| LGTGUPPUIQQLBP-ZCMDIHMWSA-N | |
| 56842015 | |
| CC(C(=O)O)NSC1=CC=CC=C1[N+](=O)[O-].C1CCC(CC1)NC2CCCCC2 |
| C21H33N3O4S | |
| MFCD00066085 | |
| N-(2-Nitrophenylthio)-L-alanine Dicyclohexylammonium Salt, N-Nps-L-alanine Dicyclohexylammonium Salt, Nps-Ala-OH.DCHA | |
| N-cyclohexylcyclohexanamine;(2S)-2-[(2-nitrophenyl)sulfanylamino]propanoic acid |
Safety and Handling
TSCA : No