missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Methoxyethyl)isopropylamine 98.0+%, TCI America™

Catalog No. p-7132479
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09875ML 5 mL
M098725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09875ML Supplier TCI America Supplier No. M09875ML

Chemical Identifiers

CAS 104678-18-4
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00144828
InChI Key UMCVTLHNNUEZDO-UHFFFAOYSA-N
PubChem CID 551575
IUPAC Name N-(2-methoxyethyl)propan-2-amine
SMILES CC(C)NCCOC

Specifications

Color Colorless
Boiling Point 125°C
Quantity 5 mL
UN Number 2734
Formula Weight 117.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Methoxyethyl)isopropylamine