missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxypropyl)morpholine 98.0+%, TCI America™

Catalog No. H043725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H043725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H043725ML Supplier TCI America Supplier No. H043725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2109-66-2
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.202
MDL Number MFCD00023378
InChI Key YAXQOLYGKLGQKA-UHFFFAOYSA-N
Synonym n-2-hydroxypropyl morpholine, 1-morpholin-4-yl propan-2-ol, 1-morpholinopropan-2-ol, 1-morpholino-2-propanol, 4-2-hydroxypropyl morpholine, 1-4-morpholino-2-propanol, 4-morpholineethanol, alpha-methyl, 4-morpholineethanol, .alpha.-methyl, acmc-1ck5f, 1-morpholino propan-2-ol
PubChem CID 97957
IUPAC Name 1-morpholin-4-ylpropan-2-ol
SMILES CC(CN1CCOCC1)O

Specifications

Color Yellow
Boiling Point 218°C
Quantity 25 mL
Formula Weight 145.20
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxypropyl)morpholine