missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)succinimide, 95%

Catalog No. AAL1130606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1130606 5 g
AAL1130614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1130606 Supplier Thermo Scientific Chemicals Supplier No. L1130606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18190-44-8 | C6H9NO3 | 143.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18190-44-8
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD00078332
InChI Key TWYIPMITVXPNEM-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl succinimide, 1-2-hydroxyethyl pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-2-hydroxyethyl, 1-2-hydroxyethyl-2,5-pyrrolidinedione, 2,5-pyrrolidinedione,1-2-hydroxyethyl, acmc-20akck, 2-hydroxyethylsuccinimide
PubChem CID 236334
IUPAC Name 1-(2-hydroxyethyl)pyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)CCO

Specifications

Melting Point 59°C to 61°C
Quantity 5 g
Beilstein 124744
Formula Weight 143.14
Percent Purity 95%
Chemical Name or Material N-(2-Hydroxyethyl)succinimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only