missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)pyrrolidine, 97%

Catalog No. AAH60368AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60368AE 100 mL
AAH60368AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60368AE Supplier Thermo Scientific Chemicals Supplier No. H60368AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2955-88-6 | C6H13NO | 115.18 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs and is also used in medicine.

Solubility
Partly soluble in water. Soluble in alcohol, ether, benzene.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-pyrrolidin-1-ylethanol
SMILES OCCN1CCCC1

Specifications

Density 0.985
Boiling Point 79°C to 81°C
Flash Point 56°C (133°F)
Refractive Index 1.471
Quantity 100 mL
UN Number UN1993
Beilstein 102976
Solubility Information Partly soluble in water. Soluble in alcohol,ether,benzene.
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material N-(2-Hydroxyethyl)pyrrolidine
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 220-976-3
RTECSNumber UY1175000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only