missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)pyrrolidine, 95%

Catalog No. p-3778794
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121200250 25 g Glass bottle
AC121201000 100 g Glass bottle
AC121205000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121200250 Supplier Thermo Scientific Chemicals Supplier No. 121200250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2955-88-6 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2955-88-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD00003181
InChI Key XBRDBODLCHKXHI-UHFFFAOYSA-N
Synonym 1-pyrrolidineethanol, n-2-hydroxyethyl pyrrolidine, 1-2-hydroxyethyl pyrrolidine, 2-pyrrolidin-1-yl ethanol, epolamine, 2-pyrrolidinoethanol, pyrrolidinoethanol, hydroxyethylpyrrolidine, 2-1-pyrrolidinyl ethanol, 2-pyrrolidin-1-yl ethan-1-ol
PubChem CID 76288
ChEBI CHEBI:48293
IUPAC Name 2-pyrrolidin-1-ylethanol
SMILES OCCN1CCCC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown to Yellow
Density 0.9800g/mL
Boiling Point 79°C to 81°C (13.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4713 to 1.4743
Quantity 25 g
Beilstein 20, II, 5
Specific Gravity 0.98
Formula Weight 115.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)pyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 220-976-3

RUO – Research Use Only