missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)phthalimide 98.0+%, TCI America™

Catalog No. p-7130437
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H024525G 25 g
H0245500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H024525G Supplier TCI America Supplier No. H024525G
Only null left

Chemical Identifiers

CAS 3891-07-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005903
InChI Key MWFLUYFYHANMCM-UHFFFAOYSA-N
PubChem CID 77499
IUPAC Name 2-(2-hydroxyethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCO

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 25 g
Formula Weight 191.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Hydroxyethyl)phthalimide
EINECSNumber (5)-0082
TSCA Yes