missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)hexamethyleneimine, 95%

CAS: 20603-00-3 | C8H17NO | 143.23 g/mol

Supplier:  Thermo Scientific Chemicals L0649714

 View more versions of this product

Catalog No. AAL0649714

Only null left
Explore available promotions

Description

Description

N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

Solubility
Fully miscible in water.

Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
Specifications

Specifications

N-(2-Hydroxyethyl)hexamethyleneimine
0.971
C8H17NO
MFCD00020988
UN2735
Air Sensitive
Fully miscible in water.
C1CCCN(CC1)CCO
143.23
143.22
20603-00-3
114°C to 115°C (23 mmHg)
1.484
25 g
104110
2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine
VMRYMOMQCYSPHS-UHFFFAOYSA-N
2-(azepan-1-yl)ethanol
88615
95%

Chemical Identifiers

20603-00-3
143.23
VMRYMOMQCYSPHS-UHFFFAOYSA-N
88615
C1CCCN(CC1)CCO
C8H17NO
MFCD00020988
2-azepan-1-yl ethanol, hexahydro-1h-azepine-1-ethanol, n-2-hydroxyethyl hexamethyleneimine, 2-azepan-1-yl ethan-1-ol, 2-hexamethyleneimino ethanol, 2-azaperhydroepinylethan-1-ol, 2-azepan-1-ylethan-1-ol, 2-azepan-1-yl ;ethanol, 2-n-hexamethyleneimino ethanol, 1-2-hydroxyethyl homopiperidine
2-(azepan-1-yl)ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 243-915-2

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only