missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. H1262500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1262500G 500 g
H126225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1262500G Supplier TCI America Supplier No. H1262500G

Chemical Identifiers

CAS 7646-67-5
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
InChI Key UUORTJUPDJJXST-UHFFFAOYSA-N
PubChem CID 193628
IUPAC Name N-(2-hydroxyethyl)prop-2-enamide
SMILES C=CC(=O)NCCO

Specifications

Boiling Point 130°C
Quantity 500 g
Formula Weight 115.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(2-Hydroxyethyl)acrylamide (stabilized with MEHQ)
EINECSNumber (2)-1023
TSCA No
Recommended Storage Refrigerator