missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Fluorophenyl)-3-oxobutyramide 98.0+%, TCI America™

Catalog No. p-7129999
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
F0963200MG 200 mg
F09631G 1 g
2 options

Catalog No. F0963200MG

Supplier: TCI America F0963200MG

Only null left

Chemical Identifiers

CAS 5279-85-6
Molecular Formula C10H10FNO2
Molecular Weight (g/mol) 195.19
MDL Number MFCD00661533
InChI Key SNNJOLBZQNBODQ-UHFFFAOYSA-N
Synonym 2′C-Fluoroacetoacetanilide
PubChem CID 763281
IUPAC Name N-(2-fluorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1F

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 200 mg
Formula Weight 195.19
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-(2-Fluorophenyl)-3-oxobutyramide