missing translation for 'onlineSavingsMsg'
Learn More

N,2'-Dimethylformanilide 97.0+%, TCI America™

Catalog No. d33681g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33681G 1 g
D33685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33681G Supplier TCI America Supplier No. D33681G

Chemical Identifiers

CAS 131840-54-5
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD09038514
InChI Key WWWSJNJIEHCOSE-UHFFFAOYSA-N
Synonym N-Methyl-N-(o-tolyl)formamide
PubChem CID 15739199
IUPAC Name N-methyl-N-(2-methylphenyl)formamide
SMILES CN(C=O)C1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 206°C
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N,2′-Dimethylformanilide