missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)morpholine 99.0+%, TCI America™

Catalog No. C118525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C118525G Supplier TCI America Supplier No. C118525G

Chemical Identifiers

CAS 4542-47-6
Molecular Formula C7H12N2O
Molecular Weight (g/mol) 140.186
MDL Number MFCD00006178
InChI Key WXVKGHVDWWXBJX-UHFFFAOYSA-N
Synonym 3-morpholinopropionitrile, 3-morpholin-4-yl propanenitrile, 3-4-morpholino propionitrile, 4-morpholinepropanenitrile, cyanoethylmorpholine, 4-morpholinepropionitrile, beta-morpholinopropionitrile, n-2-cyanoethyl morpholine, propionitrile, 3-morpholino, 3-morpholinopropanenitrile
PubChem CID 78298
IUPAC Name 3-morpholin-4-ylpropanenitrile
SMILES C1COCCN1CCC#N

Specifications

Color Yellow
Boiling Point 246°C
Quantity 25 g
Formula Weight 140.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(2-Cyanoethyl)morpholine
TSCA Yes
Recommended Storage Refrigerator