missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(2-Cyanoethyl)glycine 98.0+%, TCI America™
$65.46 - $229.10
Chemical Identifiers
| CAS | 3088-42-4 |
|---|---|
| Molecular Formula | C5H8N2O2 |
| Molecular Weight (g/mol) | 128.13 |
| MDL Number | MFCD00066903 |
| InChI Key | KZUBZCHAWPDYQX-UHFFFAOYSA-N |
| Synonym | n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid |
| PubChem CID | 76526 |
| IUPAC Name | 2-[(2-cyanoethyl)amino]acetic acid |
| SMILES | OC(=O)CNCCC#N |
Chemical Identifiers
| 3088-42-4 | |
| 128.13 | |
| KZUBZCHAWPDYQX-UHFFFAOYSA-N | |
| 76526 | |
| OC(=O)CNCCC#N |
| C5H8N2O2 | |
| MFCD00066903 | |
| n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid | |
| 2-[(2-cyanoethyl)amino]acetic acid |
Specifications
| 3088-42-4 | |
| White-Yellow | |
| MFCD00066903 | |
| n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid | |
| OC(=O)CNCCC#N | |
| 128.13 | |
| 128.13 | |
| Crystalline Powder |
| 190°C | |
| C5H8N2O2 | |
| 5 g | |
| KZUBZCHAWPDYQX-UHFFFAOYSA-N | |
| 2-[(2-cyanoethyl)amino]acetic acid | |
| 76526 | |
| ≥98.0% (T) | |
| N-(2-Cyanoethyl)glycine |
Safety and Handling
TSCA : No