missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Cyanoethyl)glycine, 98%

Catalog No. AAA1819914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1819914 25 g
AAA1819906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1819914 Supplier Thermo Scientific Chemicals Supplier No. A1819914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3088-42-4 | C5H8N2O2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3088-42-4
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00066903
InChI Key KZUBZCHAWPDYQX-UHFFFAOYSA-N
Synonym n-2-cyanoethyl glycine, glycine, n-2-cyanoethyl, n-2-cyanoethyl aminoacetic acid, 2-2-cyanoethyl amino acetic acid, 2-2-cyanoethylamino acetic acid, 2-cyanoethyl amino acetic acid, wln: qv1m2cn, .alpha.-.beta.'-cyanoethyl-aminoacetic acid
PubChem CID 76526
SMILES OC(=O)CNCCC#N

Specifications

Melting Point ∼195°C (decomposition)
UN Number UN3439
Beilstein 1756230
IUPAC Name 2-[(2-cyanoethyl)amino]acetic acid
Quantity 25 g
Formula Weight 128.13
Percent Purity 98%
Chemical Name or Material N-(2-Cyanoethyl)glycine
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 221-418-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only