missing translation for 'onlineSavingsMsg'
Learn More

N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC53646DA

Quantity 1g

 View more versions of this product

Catalog No. CC53646DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

748187-76-0
156.61
NIBLXLUVASMQFM-UHFFFAOYSA-N
2794168
CNCC1=CC(Cl)=NC=C1
C7H9ClN2
MFCD06654462
n-2-chloropyridin-4-yl methyl-n-methylamine, 2-chloropyridin-4-yl methyl methyl amine, 1-2-chloropyridin-4-yl-n-methylmethanamine, 2-chloro-4-methylamino methyl pyridine, 4-pyridinemethanamine,2-chloro-n-methyl, 2-chloro 4-pyridyl methyl methylamine, 4-pyridinemethanamine, 2-chloro-n-methyl, 2-chloro-n-methyl-4-pyridinemethanamine, 2-chloro-pyridin-4-ylmethyl-methyl-amine
1-(2-chloropyridin-4-yl)-N-methylmethanamine

Specifications

N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine
96.5
97%
C7H9ClN2
MFCD06654462
NIBLXLUVASMQFM-UHFFFAOYSA-N
1-(2-chloropyridin-4-yl)-N-methylmethanamine
2794168
97%
748187-76-0
100.0
Amber glass bottle
1g
n-2-chloropyridin-4-yl methyl-n-methylamine, 2-chloropyridin-4-yl methyl methyl amine, 1-2-chloropyridin-4-yl-n-methylmethanamine, 2-chloro-4-methylamino methyl pyridine, 4-pyridinemethanamine,2-chloro-n-methyl, 2-chloro 4-pyridyl methyl methylamine, 4-pyridinemethanamine, 2-chloro-n-methyl, 2-chloro-n-methyl-4-pyridinemethanamine, 2-chloro-pyridin-4-ylmethyl-methyl-amine
CNCC1=CC(Cl)=NC=C1
156.61
156.62
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents