missing translation for 'onlineSavingsMsg'
Learn More

N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine, 97%, Thermo Scientific™

Catalog No. CC53646CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC53646CB 250mg
CC53646DA 1g
CC53646DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC53646CB Supplier Thermo Scientific Chemicals Supplier No. CC53646CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 748187-76-0
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD06654462
InChI Key NIBLXLUVASMQFM-UHFFFAOYSA-N
Synonym n-2-chloropyridin-4-yl methyl-n-methylamine, 2-chloropyridin-4-yl methyl methyl amine, 1-2-chloropyridin-4-yl-n-methylmethanamine, 2-chloro-4-methylamino methyl pyridine, 4-pyridinemethanamine,2-chloro-n-methyl, 2-chloro 4-pyridyl methyl methylamine, 4-pyridinemethanamine, 2-chloro-n-methyl, 2-chloro-n-methyl-4-pyridinemethanamine, 2-chloro-pyridin-4-ylmethyl-methyl-amine
PubChem CID 2794168
IUPAC Name 1-(2-chloropyridin-4-yl)-N-methylmethanamine
SMILES CNCC1=CC(Cl)=NC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 156.62
Percent Purity 97%
Chemical Name or Material N-[(2-Chloropyridin-4-yl)methyl]-N-methylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.