Learn More
N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%
CAS: 7250-67-1 | C6H12ClN·HCl | 170.08 g/mol
$306.11 - $306.11
Chemical Identifiers
| CAS | 7250-67-1 |
|---|---|
| Molecular Formula | C6H12ClN·HCl |
| Molecular Weight (g/mol) | 170.08 |
| MDL Number | MFCD00012718 |
| InChI Key | FSNGFFWICFYWQC-UHFFFAOYSA-N |
| Synonym | 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride |
| PubChem CID | 81668 |
| IUPAC Name | 1-(2-chloroethyl)pyrrolidine;hydrochloride |
| SMILES | C1CCN(C1)CCCl.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC109221000
|
Thermo Scientific Chemicals
109221000 |
100 g | Glass bottle |
Each for $306.11
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7250-67-1 | |
| 170.08 | |
| FSNGFFWICFYWQC-UHFFFAOYSA-N | |
| 81668 | |
| C1CCN(C1)CCCl.Cl |
| C6H12ClN·HCl | |
| MFCD00012718 | |
| 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride | |
| 1-(2-chloroethyl)pyrrolidine;hydrochloride |
Specifications
| 7250-67-1 | |
| 100.0 | |
| White | |
| 98% | |
| C6H12ClN·HCl | |
| 100 g | |
| 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride | |
| FSNGFFWICFYWQC-UHFFFAOYSA-N | |
| 1-(2-chloroethyl)pyrrolidine;hydrochloride | |
| 81668 | |
| 98% | |
| N-(2-Chloroethyl)pyrrolidine hydrochloride |
| 97.5 | |
| 171.0°C to 175.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00012718 | |
| 20, II, 4 | |
| Solubility in water: soluble | |
| C1CCN(C1)CCCl.Cl | |
| 170.08 | |
| 170.08 | |
| Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
GHS Signal Word: Warning
EINECSNumber : 230-660-7
RUO – Research Use Only