missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%

Catalog No. AC109220250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109220250 25 g Glass bottle
AC109221000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109220250 Supplier Thermo Scientific Chemicals Supplier No. 109220250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7250-67-1 | C6H12ClN·HCl | 170.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7250-67-1
Molecular Formula C6H12ClN·HCl
Molecular Weight (g/mol) 170.08
MDL Number MFCD00012718
InChI Key FSNGFFWICFYWQC-UHFFFAOYSA-N
Synonym 1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
PubChem CID 81668
IUPAC Name 1-(2-chloroethyl)pyrrolidine;hydrochloride
SMILES C1CCN(C1)CCCl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 20, II, 4
Solubility Information Solubility in water: soluble
Formula Weight 170.08
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-(2-Chloroethyl)pyrrolidine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
EINECSNumber 230-660-7

RUO – Research Use Only