missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Bromoethyl)phthalimide, 97+%

Catalog No. AAA1216414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1216414 25 g
AAA1216422 100 g
AAA1216436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1216414 Supplier Thermo Scientific Chemicals Supplier No. A1216414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 574-98-1 | C10H8BrNO2 | 254.083 g/mol

N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Bromoethyl)phthalimide is an intermediate used in organic synthesis. It can react with phenyl magnesium bromide to get 2-(2-bromo-ethyl)-3-hydroxy-3-phenyl-isoindolin-1-one.

Solubility
Soluble in water (224 mg/L at 25°C).

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 574-98-1
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
MDL Number MFCD00005902
InChI Key CHZXTOCAICMPQR-UHFFFAOYSA-N
Synonym n-2-bromoethyl phthalimide, 2-2-bromoethyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-bromoethyl, 2-2-bromoethyl-1h-isoindole-1,3 2h-dione, beta-bromoethylphthalimide, 2-bromoethyl phthalimide, 1-bromo-2-phthalimidoethane, beta-phthalimidoethyl bromide, n-2-bromoethyl-phthalimide, 2-2-bromo-ethyl-isoindole-1,3-dione
PubChem CID 11325
IUPAC Name 2-(2-bromoethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCBr

Specifications

Melting Point 80°C to 84°C
Quantity 25 g
Beilstein 148736
Solubility Information Soluble in water (224mg/L at 25°C).
Formula Weight 254.09
Percent Purity ≥98%
Chemical Name or Material N- (2-Bromoethyl)phthalimide
EINECSNumber 209-379-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only